Skip to Content
Merck
All Photos(1)

Key Documents

Z281298

Laboratory brushes

Cylinder, L 20 in., brush diam. × L 2 3/8 in. × 6 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131605
NACRES:
NB.35
Pricing and availability is not currently available.

packaging

pkg of 4 ea

L

20 in.

brush diam. × L

2 3/8 in. × 6 in.

Looking for similar products? Visit Product Comparison Guide

General description

Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service