Skip to Content
Merck
All Photos(1)

Documents

T25801

Sigma-Aldrich

Tetraphenylcyclopentadienone

98%

Synonym(s):

2,3,4,5-Tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-Tetraphenylcyclopenta-2,4-dienone, 2,3,4,5-Tetraphenylcyclopentadienone, Cyclone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H20O
CAS Number:
Molecular Weight:
384.47
Beilstein:
683115
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

217-220 °C (lit.)

SMILES string

O=C1C(c2ccccc2)=C(c3ccccc3)C(c4ccccc4)=C1c5ccccc5

InChI

1S/C29H20O/c30-29-27(23-17-9-3-10-18-23)25(21-13-5-1-6-14-21)26(22-15-7-2-8-16-22)28(29)24-19-11-4-12-20-24/h1-20H

InChI key

PLGPSDNOLCVGSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Kenny et al.
The Annals of occupational hygiene, 45(1), 35-42 (2001-01-04)
Porous polyurethane foams provide a low-cost method for separating dust into health-related fractions in accordance with recognized sampling conventions. A number of dust sampling instruments that make use of foam selectors have been described in recent literature, but practical experiences
Ingo Oesterling et al.
Journal of the American Chemical Society, 129(15), 4595-4605 (2007-03-24)
Two routes for the introduction of highly fluorescent peryleneimide chromophores into the scaffolding of polyphenylene dendrimers via iterative Diels-Alder cycloadditions are presented. The key intermediates for the divergent dendrimer buildup were two cyclopentadienone branching units carrying two peryleneimides and two
Ming-Yi Leung et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(39), 14013-14021 (2016-08-10)
A new class of ruthenium(II) polypyridine complexes with a series of D-π-A-π-D type (D=donor, A=acceptor) ligands was synthesized and characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service