Key Documents
SYNB300250C
Synthware gas washing bottle with fritted disc
250 mL, joint: ST/NS 45/50, Disc porosity C
Synonym(s):
Synthware B300250C, Drechsel bottle, gas washing bottle
About This Item
UNSPSC Code:
41103209
NACRES:
NB.51
material
clear bottle
cylindrical bottle
straight bottle
Joint
joint: ST/NS45/50
feature
cap
packaging
pkg of 1 ea
manufacturer/tradename
Synthware B300250C
H
305 mm
capacity
250 mL
250 mL
Related Categories
General description
Gas washing bottle with standard taper 45/50 cap style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material. Available with a coarse or extra coarse fritted disc. Order plastic clip separately. (not supplied)
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)