Skip to Content
Merck
All Photos(2)

Key Documents

N1280

Sigma-Aldrich

Norclomipramine hydrochloride

≥98% (TLC), powder

Synonym(s):

N-Desmethylclomipramine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21ClN2 · HCl
CAS Number:
Molecular Weight:
337.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to yellow

SMILES string

Cl.CNCCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C18H21ClN2.ClH/c1-20-11-4-12-21-17-6-3-2-5-14(17)7-8-15-9-10-16(19)13-18(15)21;/h2-3,5-6,9-10,13,20H,4,7-8,11-12H2,1H3;1H

InChI key

KMDDAZOLOSKTKZ-UHFFFAOYSA-N

Biochem/physiol Actions

Tricyclic antidepressant; an active metabolite of clomipramine that preferentially blocks the norepinephrine transporter.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Fujita et al.
European journal of pharmacology, 204(3), 227-233 (1991-11-12)
When rats received a single i.p. injection of clomipramine (20 mg/kg), clomipramine and desmethylclomipramine were rapidly distributed into the brain and their concentrations were markedly higher in the brain than in the serum, while the concentration of the metabolite in
S Härtter et al.
Psychopharmacology, 117(2), 149-153 (1995-01-01)
The selective serotonin reuptake inhibitor fluvoxamine interferes with the metabolism of tricyclic antidepressants. The present investigation was set out to characterize these interactions in vitro using rat liver microsomes and in vivo by analysing levels of clomipramine and metabolites in
J N King et al.
American journal of veterinary research, 61(1), 80-85 (2000-01-12)
To determine pharmacokinetics of clomipramine and its principle metabolite (desmethylclomipramine) in the plasma of dogs following single-dose and repeated-dose oral administration at various dosages. 9 male and 9 female Beagles. Clomipramine was administered orally at a dose of 1, 2
O Dale et al.
Veterinary and human toxicology, 36(4), 309-310 (1994-08-01)
A 27-y-old male was admitted deeply comatosed 5-6 h after taking approximately 15 g clomipramine. The prominent feature of the case was a biphasic course of clomipramine and desmethylclomipramine serum concentrations, possibly caused by delayed drug absorption. Clinically, 2 serious
Plasma concentrations after a clomipramine intoxication.
L M Stolk et al.
Journal of analytical toxicology, 22(7), 612-613 (1998-12-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service