M31651
4-Methylbenzyl cyanide
98%
Synonym(s):
p-Tolylacetonitrile, α-Cyano-p-xylene, 4-Methylbenzyl cyanide, 4-Methylphenylacetonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H4CH2CN
CAS Number:
Molecular Weight:
131.17
Beilstein:
1099645
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.519 (lit.)
bp
242-243 °C (lit.)
mp
18 °C (lit.)
density
0.992 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(CC#N)cc1
InChI
1S/C9H9N/c1-8-2-4-9(5-3-8)6-7-10/h2-5H,6H2,1H3
InChI key
RNHKXHKUKJXLAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
222.8 °F - closed cup
Flash Point(C)
106 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jun-Nan Yang et al.
Insect science, 28(3), 811-824 (2020-05-21)
The pea leafminer, Liriomyza huidobrensis, is an important pest species affecting ornamental crops worldwide. Plant damage consists of oviposition and feeding punctures created by female adult flies as well as larva-bored mines in leaf mesophyll tissues. How plants indirectly defend
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service