Skip to Content
Merck
All Photos(1)

Key Documents

L040031

Sigma-Aldrich

Cap A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

concentration

97.0-119.0 g/L (acetic anhydride, titration)

color

clear colorlessto brownish

Looking for similar products? Visit Product Comparison Guide

General description

Cap A solution consists of acetic anhydrideanhydride, lutidine, and tetrahydrofuran (THF).

Application

Cap A solution is used after the phosphoramidite reaction/coupling step.

Biochem/physiol Actions

Cap A solution aborts chains bearing unreacted 5′ hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′ hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service