Skip to Content
Merck
All Photos(4)

Key Documents

F300

Sigma-Aldrich

Iron(III) acetylacetonate

greener alternative

97%

Synonym(s):

2,4-Pentanedione iron(III) derivative, Fe(acac)3, Ferric acetylacetonate, Iron(III) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(C5H7O2)3
CAS Number:
Molecular Weight:
353.17
Beilstein:
4157960
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

reaction suitability

core: iron
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

180-182 °C (dec.) (lit.)

density

5.24 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CC(=O)\C=C(\C)O[Fe](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

AQBLLJNPHDIAPN-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.
Non-Precious metal catalyst used by Baran in 2014 to affect the complexity generating reductive coupling of olefins

A Practical and Catalytic Reductive Olefin Coupling

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukáš Pečinka et al.
Rapid communications in mass spectrometry : RCM, 34(11), e8749-e8749 (2020-02-13)
Gold-iron bimetallic materials have applications in many fields, especially in nanotechnology and biomedicine. The chemistry of iron-doped gold clusters is still not fully understood but opens up the possibility of developing new materials, e.g. of gold cages doped with iron
Ruth Rodríguez-Ramos et al.
Chemosphere, 236, 124377-124377 (2019-09-25)
In this study, the application of different nanomaterials as dispersants in matrix solid phase dispersion has been evaluated for the extraction of fifteen phthalates from different environmental samples prior to their separation and quantification by ultra-high performance liquid chromatography coupled
Roland Schoch et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(17), 3768-3775 (2014-09-13)
A new iron-based catalyst for carbon monoxide oxidation, as a potential substitute for precious-metal systems, has been prepared by using a facile impregnation method with iron tris-acetylacetonate as a precursor on γ-Al2 O3 . Light-off and full conversion temperatures as
Tingting Li et al.
Nanoscale, 7(9), 4171-4178 (2015-02-12)
The excellent electrochemical performance of greigite (Fe3S4) coupled with its vast abundance and low toxicity make it a good prospect as an anode material for lithium ion batteries (LIBs). In this research, a simple and feasible approach for producing pure
Steffen Preissler et al.
eLife, 9 (2020-12-10)
The metazoan endoplasmic reticulum (ER) serves both as a hub for maturation of secreted proteins and as an intracellular calcium storage compartment, facilitating calcium-release-dependent cellular processes. ER calcium depletion robustly activates the unfolded protein response (UPR). However, it is unclear

Articles

Metal-organic frameworks (MOFs) are porous, crystalline materials composed of metal ions bound by organic ligands.

Metal-organic frameworks (MOFs) are porous, crystalline materials composed of metal ions bound by organic ligands.

Metal-organic frameworks (MOFs) are porous, crystalline materials composed of metal ions bound by organic ligands.

Metal-organic frameworks (MOFs) are porous, crystalline materials composed of metal ions bound by organic ligands.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service