DWKBWP1000P
Azlon Polypropylene Wide Neck Round Bottles with Screw Cap
round translucent polypropylene bottle, capacity 1000 mL
Synonym(s):
BWP1000P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121818
NACRES:
NB.43
Recommended Products
material
round translucent polypropylene bottle
wide-neck
sterility
non-sterile
packaging
pkg of 5 ea
manufacturer/tradename
Azlon BWP1000P
capacity
1000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Featuring a leakproof, linerless cap design, AZLON® wide neck polypropylene bottles are easy to fill and autoclavable at 121°C.
Application
Ideal for a wide range of liquid storage and sampling applications
Features and Benefits
Autoclavable at 121°C and suitable for food contact applications
Legal Information
AZLON is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service