DWK257105451
DURAN® filtration apparatus complete for slit-sleeve disc with PTFE insert
clamp, filter flask and funnel, filter thickness (47 mm), non-sterile
Synonym(s):
257105451
All Photos(2)
About This Item
UNSPSC Code:
41104925
NACRES:
NB.17
Recommended Products
material
borosilicate glass 3.3 filter
sterility
non-sterile
packaging
case of 1 ea
manufacturer/tradename
DURAN® 257105451
filter thickness
(47 mm)
General description
Virtually universal applications, as the medium only comes into contact with glass and PTFE. Fired-on, highly durable white print. Replaceable plates and PTFE adapters allow easy and fast cleaning from both sides. The tried-and-tested DURAN filtering flask and PTFE hose connection (suitable for tubes with ID 8 mm) enable safe working in the laboratory. The replaceable plates and the PTFE adapter in conjunction with the clamp enable rapid changing of porosities or replacement of filters. Easy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white print.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)