Skip to Content
Merck
All Photos(1)

Key Documents

DWK231885404

Reagent Bottle, Wide Neck

wide-neck

Synonym(s):

231885404

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

amber soda-lime glass bottle
wide-neck

Joint

Joints ST/NS neck WITH STOPPER (NS 60/46)

packaging

pkg of 10 ea

manufacturer/tradename

231885404

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

Application

Media preparation and cultivation of light-sensitive media

Features and Benefits

A maximum usage temperature of + 100 °C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3., If the bottle and the stopper have a temperature difference greater than 30 °C, the glass stoppers can become stuck, Easy access to the bottle contents due to the wide diameter of the bottle neck ;

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service