BR781344
BRAND® strips of 8 caps
polypropylene, blue, for PCR tubes
Synonym(s):
PCR, PCR Tubes, Tube Strips, Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
blue
domed
polypropylene
feature
for PCR tubes
packaging
pack of 125 ea
pack of 125 ea
manufacturer/tradename
BRAND 781344
technique(s)
PCR: suitable
Looking for similar products? Visit Product Comparison Guide
General description
With PCR cap strips you can seal PCR strips and plates with 24, 48, and 96 wells reliably and protect your samples effectively against evaporation and contamination.The shape of the cap strips is optimally matched to BRAND PCR tube strips and plates so they are easy to open and close. To prevent contamination, the cap strips are equipped with two side tabs for exact positioning before closing and a small opening attachment on each cap. This prevents contact with the inside of the cap.The highly transparent flat cap strips are ideal for qPCR because the cap shape allows accurate fluorescence measurement. With domed cap strips, thermal cyclers without pressure sensors exert a stronger force, thus further increasing the sealing effect.Manufactured under cleanroom conditions. BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service