BR780540
BRAND® disposable microcentrifuge tube
with safety lid, capacity 1.5 mL
Synonym(s):
Microcentrifuge Tubes, Tubes
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
clear polypropylene
sterility
non-sterile
feature
with safety lid
packaging
pack of 1000
manufacturer/tradename
BRAND 780540
parameter
30000 × g max. RCF
capacity
1.5 mL
suitability
suitable for (Free of endotoxins, DNase and Rnase)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
DNA isolation and preparation of the master mix are essential for a successful PCR, require the highest possible product cleanliness, good impermeability, and the option for use in high throughput analysis. BRAND offers microtubes (microcentrifuge tubes) that fulfill all the requirements for use in the PCR applications.
Application
PCR suitable microtubes find applications in areas related to:
- DNA and RNA isolation
- DNA and RNA purification
- Preparing a master mix
- Sample dilution
Features and Benefits
- The lid closure on the microcentrifuge tube protects against accidental opening of the lid.
- The wide lip on the cap allows for one-handed handling.
- Tight-sealing cap.
- Free from PCR inhibitors.
- Highly transparent and consistent wall thickness.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service