BR148502
BRAND® funnel holder
Holds 2 funnels
Synonym(s):
Accessories, Funnels
All Photos(2)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.35
Recommended Products
material
polypropylene
feature
Holds 2 funnels
packaging
pack of 5 ea
manufacturer/tradename
BRAND 148502
Related Categories
General description
PP. To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8-14 mm with a locking nut
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)