Key Documents
BR140006
BRAND® separating funnel with screw cap
size 500 mL
Synonym(s):
Funnels, Separating funnels
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
conical PTFE stopcock
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
BRAND 140006
size
500 mL
Related Categories
General description
Polypropylene, high clarity for observation of contents. With polypropylene screw cap and special conical PTFE stopcock with retention device. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)