Skip to Content
Merck
All Photos(1)

Documents

938211

Sigma-Aldrich

Manganese ferrite nanoparticles

new

silica coated, 100 nm particle size, 0.1 wt. % in water

Synonym(s):

dispersed silica-coated manganese ferrite nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303

Quality Level

form

dispersion

concentration

0.1 wt. % in water

particle size

100 nm

bp

>1000 °C (Manganese ferrite)

mp

>300 °C (Manganese ferrite)

SMILES string

[O].[Mn].[Fe]

InChI

1S/Fe.Mn.O

General description

Silica coated manganese ferrite nanoparticles, 100nm particle size, 0.1 wt% in water, is designed by coating a silica shell on Manganese ferrite nanoparticles (NPs), enabling the water-solubility and biocompatibility of the NPs. The coating provides a protective and functional surface layer to the manganese ferrite nanoparticles, offering various advantages and potential applications in research work.

Application

Silica coated manganese ferrite nanoparticles find applications in the research areas of targeted drug delivery and magnetic resonance imaging (MRI) applications. The particles exhibit superparamagnetic behavior suitable for MRI contrast agents. In a study, silica-coated manganese ferrite nanoparticles has been found as a highly efficient T2 contrast agent compared to other reported nanoparticles justifying their potential for MRI based diagnosis of cancer. Studies found Silica coated manganese ferrite nanoparticles to be efficient, selective, and reusable adsorbents for methylene blue (MB) removal, which is a pollutant implicated in health issues, such as eye irritation and itchy skin as well as respiratory and digestive problems and is commonly used as a dye in the textile industry, for tissue staining, and in the leather tanning industry.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly stable silica-coated manganese ferrite nanoparticles as high-efficacy T2 contrast agents for magnetic resonance imaging
Ashfaq Ahmad, et al.
AIP Advances, 8 (2018)
Silica-coated magnesium ferrite nano adsorbent for selective removal of methylene blue
Supawitch Hoijang, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 606, 125483-125483 (2020)
Biocompatible mesoporous silica-coated superparamagnetic manganese ferrite nanoparticles for targeted drug delivery and MR imaging applications
B Sahoo, et al.
Journal of Colloid and Interface Science, 431, 31-41 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service