Skip to Content
Merck
All Photos(1)

Key Documents

933724

Sigma-Aldrich

Linear polyethylenimine-block-poly(ethylene glycol)

PEG average Mn 5,000, PEI average Mn 10,000

Synonym(s):

PEG-PEI, PEG-b-PEI, Poly(ethylene glycol)-block-polyethylenimine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C2H4O)n(C2H5N)mOH
UNSPSC Code:
12352207
NACRES:
NA.21

form

powder or solid

Quality Level

mol wt

PEG Mn 5,000 Da
PEG average Mn 5,000
PEI Mn 10,000 Da
PEI average Mn 10,000

color

white to pale yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Due to their low immunogenicity profiles, polymeric nonviral vectors have shown promise in advancing the treatment of many severe genetic and acquired diseases via gene therapy. Poly(ethyleneimine) is one of the most commonly used cationic polymers for gene delivery applications. Copolymers of PEI and hydrophilic poly(ethylene glycol) have been designed to provide a stealth-like shield around polymer/DNA complexes, reducing nonspecific interactions, inhibiting activation of the reticuloendothelial system, and prolonging the half-life of the complexes in the blood.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Brissault et al.
Biomacromolecules, 7(10), 2863-2870 (2006-10-10)
The study of ethyloxazoline/methyloxazoline (EtOXZ/MeOXZ) copolymerization, initiated by methyl tosylate (MeOTs), showed that (i) incorporation of MeOXZ units into random copolymer becomes effective over DP = 100 and (ii) propagation process proceeds with negligible transfer to monomer up to a
Pallab Banerjee et al.
Bioconjugate chemistry, 17(1), 125-131 (2006-01-19)
A block copolymer of a hyperbranched poly(ethylene glycol)-like core and linear polyethylenimine (HBP) was synthesized by a facile synthetic route that included (1) a single-step cationic copolymerization of diepoxy and polyhydroxyl monomers, (2) derivatization of hydroxyl groups of the core
Dagmar Fischer et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(9), 983-992 (2004-08-21)
The in vivo body distribution and the pharmacokinetics of a 20mer double-stranded nuclear factor kappaB decoy oligodeoxynucleotide (ODN) complexed with 25-kDa poly(ethylene imine) (PEI), low molecular weight 2.7-kDa PEI, and PEGylated PEI [bPEI(25k)-glPEG(550)(50)] after intravenous injection were studied in BALB/c

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service