932272
Poly[(9,9-dioctylfluorenyl-2,7-diyl)-co-(2,5-p -xylene)]
Synonym(s):
ADS145UV, PFO-co-pX, PFP
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
powder
solid (powder, crystal or fiber)
Quality Level
mol wt
Mw >20,000 Da by GPC
color
white to off-white
solubility
THF: soluble
chlorobenzene: soluble
chloroform: soluble
dichlorobenzene: soluble
toluene: soluble
λmax
335 nm±10 nm in THF
fluorescence
λem 378 nm±10 nm in THF
Related Categories
Application
Polymer, Emitting Layers for Solution-Processed OLEDs; blue emission.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Materials (Basel, Switzerland), 10(3) (2017-08-05)
The optical of conjugated polymer poly[(9,9-dioctylfluorenyl-2,7-diyl)-co-(2,5-p-xylene)] also known as [(PFO-co-pX), ADS145UV], dissolved in a few solvents, has been measured. The absorption, emission spectra, and quantum yield have been investigated by using a spectrophotometer and spectrofluorometer, respectively. The properties of Amplified
Photophysical Probing of Dye Microenvironment, Diffusion Dynamics and Energy Transfer.
The Journal of Physical Chemistry C, 122, 6900?6911-6900?6911 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service