Skip to Content
Merck
All Photos(1)

Key Documents

920703

Sigma-Aldrich

((1-(tert-Butoxycarbonyl)piperidin-4-yl)methyl)proline

Synonym(s):

Rigid linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N2O4
Molecular Weight:
312.40
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

CC(C)(OC(N1CCC(CC1)CN2CCCC2C(O)=O)=O)C

InChI

1S/C16H28N2O4/c1-16(2,3)22-15(21)17-9-6-12(7-10-17)11-18-8-4-5-13(18)14(19)20/h12-13H,4-11H2,1-3H3,(H,19,20)

InChI key

RIWULNINLIEMRM-UHFFFAOYSA-N

Application

((1-(tert-Butoxycarbonyl)piperidin-4-yl)methyl)proline is useful as a rigid linker in PROTAC development for targeted protein degradation as well as chemical conjugates. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties. Developed in partnership with ComInnex.

Similar linkers in this series:
920533
920649
920665
920673

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service