916838
D-Lactic acid-1-13C
≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity)
About This Item
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥98% (CP)
≥98% (Chiral Purity)
form
solid
greener alternative product score
old score: 45
new score: 14
Find out more about DOZN™ Scoring
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
mass shift
M+1
storage temp.
−20°C
SMILES string
O[13C](=O)[C@H](O)C
InChI
1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m1/s1/i3+1
InChI key
JVTAAEKCZFNVCJ-DFXGSJBSSA-N
Related Categories
General description
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1C
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service