All Photos(1)
About This Item
Empirical Formula (Hill Notation):
WS2
CAS Number:
Molecular Weight:
247.97
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
Looking for similar products? Visit Product Comparison Guide
General description
The transformation of 2D materials in QDs form supplement excellent properties from their quantum confinement effects, surface-volume ratio, solubility, edge-effects, tunable PL, ease to functionalize and fluorescence properties. These 2D QDs are demanding to impart new dimension to the 2D nanosheets and in order to advance to next level, the better understanding of the properties, synthesis methods and applications of 2D QDs are needed. The exciting optical properties of the WS2 QDs such as strong excitonic absorption bands covering the visible region, excellent excitonic fluorescence, highly tunable PL, atomically-thin structure, molecular size, biocompatibility, easy functionalization, enhanced photostability, higher molar extinction coefficients and long-term photostability make them attractive candidates for optical sensing.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
2D−Materials-Based Quantum Dots: Gateway Towards Next-Generation Optical Devices
S. Dhanabalan
Advanced Optical Materials, 1700257-1700257 (2017)
Emerging 0D Transition-Metal Dichalcogenides for Sensors, Biomedicine, and Clean Energy
B. Li
Small, 13, 1700527-1700527 (2017)
Research Update: Beyond graphene?Synthesis of functionalized quantum dots of 2D materials and their applications
Musselman, et al.
APL Materials, 6, 120701-120701 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service