Skip to Content
Merck
All Photos(1)

Key Documents

901971

Sigma-Aldrich

SunTronic® silver nanoparticle ink for inkjet printing

sintering temperature: 150-250 °C

Synonym(s):

Ag ink, Conductive ink, Silver ink, SunTronic®EMD5730

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352103
NACRES:
NA.23

description

Sintering temperature: 150-250 °C for 10 – 60 min
Solid content: 40%
Volume Resistivity: 5 – 30 uΩ.cm

form

liquid

surface tension

28-31 dyn/cm

viscosity

11-13 cP (Brookfield)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

SunTronic EMD5730 is formulated for inkjet printing of conductive traces onto a variety of substrates. EMD5730 is a solvent-based silver nanoparticle ink which demonstrates superior conductivity, stable jetting performance and adhesion to ITO coated glass.
EMD5730 can be used in the manufacture of touch panel displays, printed RFID antennae, smart cards, sensors, security labels, touch switches and other printed electronics applications.

Application

SunTronic® silver nanoparticle ink for inkjet printing (SunTronic® EMD5730) is a 3D additive printing technology that is used in a variety of applications such as flexible microsystems, and electronics. It is printed on a liquid crystal polymeric substrate for the formation of a low cost, adaptable tooling for effective deposition of a wide range of materials.

Caution

Printing
EMD5730 is supplied as ready-to-use solvent based inks, and can be printed on various deposition printers and jetted on through various piezo DOD inkjet printheads suitable for solvent based inks, although full printhead compatibility will need to be verified in each case.
Sintering:
Sintering conditions may change for different ink film thicknesses. Typical sintering temperature may range from 150-250°C depending on temperature tolerance of the substrate. Sintering time may range from 10-30 minutes. Electrical Resistance may be reduced with higher sintering temperature and/or longer sintering times.
Storage at a cool room temperature 10-15°C is recommended. Refrigeration can be used but agitated to assure proper dispersion is recommended.

Legal Information

SunTronic is a registered trademark of Sun Chemical, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Low-Temperature Assembly of Surface-Mount Device on Flexible Substrate Using Additive Printing Process
2018 IEEE 68th Electronic Components and Technology Conference, 379-385 (2018)
Study of inkjet-printed serpentine structure on flexible substrates deformed over sculptured surfaces
Bower IA, et al.
Flexible and Printed Electronics (2020)
Additively Manufactured mm-Wave Multichip Modules With Fully Printed``Smart''Encapsulation Structures
He X, et al.
IEEE Transactions on Components, Hybrids, and Manufacturing (2019)

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service