Skip to Content
Merck
All Photos(1)

Key Documents

901466

Sigma-Aldrich

Mes-Umemoto reagent

≥95%

Synonym(s):

Dimesityl(trifluoromethyl)sulfonium trifluoromethanesulfonate, MacMillan trifluoromethylation reagent, dMesSCF3 reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22F6O3S2
CAS Number:
Molecular Weight:
488.51
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

functional group

fluoro
thioether
triflate

storage temp.

−20°C

SMILES string

FC(F)(F)[S+](c2c(cc(cc2C)C)C)c1c(cc(cc1C)C)C.FC(F)(F)[S](=O)(=O)[O-]

InChI

1S/C19H22F3S.CHF3O3S/c1-11-7-13(3)17(14(4)8-11)23(19(20,21)22)18-15(5)9-12(2)10-16(18)6;2-1(3,4)8(5,6)7/h7-10H,1-6H3;(H,5,6,7)/q+1;/p-1

InChI key

HAYTWACXJNSBRL-UHFFFAOYSA-M

Application

Under a dual catalytic copper/photoredox manifold, the Mes-Umemoto reagent has been demonstrated by the MacMillan lab to be an excellent reagent for the trifluoromethylation of alkyl halides and aryl halides to yield alkyl-CF3 and aryl-CF3 in high yields. In both cases, these reactions exhibit wide substrate scope with good functional group tolerance. More specifically, a variety of 5-membered and 6-membered heteroaryl halides can be readily converted to the corresponding trifluoromethylheteroarenes under mild conditions. To be use in conjunction with supersilanol (902489) and Ir photocatalyst (902217 or 902225).

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shi-Meng Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(19), 6542-6546 (2016-03-08)
A Pd-catalyzed Suzuki cross-coupling of arylboronic acids with Yagupolskii-Umemoto reagents was explored. In contrary to trifluoromethylations, the Pd-catalyzed reaction of R-B(OH)2 and [Ar2 SCF3 ](+) [OTf](-) provided the arylation products (R-Ar) in good to high yields. The reaction confirms that
Chip Le et al.
Science (New York, N.Y.), 360(6392), 1010-1014 (2018-06-02)
Transition metal-catalyzed arene functionalization has been widely used for molecular synthesis over the past century. In this arena, copper catalysis has long been considered a privileged platform due to the propensity of high-valent copper to undergo reductive elimination with a

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service