Skip to Content
Merck
All Photos(1)

Key Documents

900911

L-Isoleucine-13C,d7 (2,3,4,4-d4-3-methyl-d3, 5-13C)

≥95 atom % D, ≥97 atom % 13C, ≥95% (CP), optical purity≥98% (at α-carbon)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5D7H6NO2
Molecular Weight:
139.21
UNSPSC Code:
12352209

isotopic purity

≥97 atom % 13C
≥95 atom % D

Assay

≥95% (CP)

form

solid

optical purity

≥98% (at α-carbon)

technique(s)

bio NMR: suitable
protein expression: suitable

mass shift

M+8

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service