Skip to Content
Merck
All Photos(1)

Key Documents

900593

Sigma-Aldrich

PAd-DalPhos

95%

Synonym(s):

8-[2-[Bis(2-methylphenyl)phosphino]phenyl]-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.13,7]decane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H34O3P2
CAS Number:
Molecular Weight:
504.54
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

functional group

phosphine

Related Categories

Application

PAd-DalPhos is a versatile air stable pre-catalyst for C(sp2)-N coupling. It catalyzes N-arylation of amides with (hetero)aryl (pseudo)halide. It can also be used in the synthesis of unsymmetrical 1,3-di(hetero)aryl-1H-indazoles from hydrazine, o-chloro (hetero)benzophenones and (hetero)aryl bromides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ni and Cu-catalyzed one pot synthesis of unsymmetrical 1, 3-di (hetero) aryl-1H-indazoles from hydrazine, o-chloro (hetero) benzophenones, and (hetero) aryl bromides.
Wiethan C, et al.
Organic & Biomolecular Chemistry, 15(23), 5062-5069 (2017)
Nickel?Catalyzed N?Arylation of Primary Amides and Lactams with Activated (Hetero) aryl Electrophiles.
Lavoie C M, et al.
Chemistry?A European Journal , 22(52), 18752-18755 (2016)
Christopher M Lavoie et al.
Nature communications, 7, 11073-11073 (2016-03-24)
Palladium-catalysed C(sp(2))-N cross-coupling (that is, Buchwald-Hartwig amination) is employed widely in synthetic chemistry, including in the pharmaceutical industry, for the synthesis of (hetero)aniline derivatives. However, the cost and relative scarcity of palladium provides motivation for the development of alternative, more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service