Skip to Content
Merck
All Photos(1)

Documents

809845

Sigma-Aldrich

Pregnenolone-2,2,4,4-d4

≥98 atom % D, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D4H28O2
Molecular Weight:
320.50
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

O=C(C)[C@H]1CCC2C3CC=C4C([2H])([2H])[C@@H](O)C([2H])([2H])C[C@]4(C)C3CC[C@@]21C

InChI

1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16?,17+,18?,19?,20-,21+/m0/s1/i8D2,12D2

InChI key

ORNBQBCIOKFOEO-JHAWGCOXSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service