Skip to Content
Merck
All Photos(1)

Key Documents

796948

Sigma-Aldrich

Sodium selenide

95%

Synonym(s):

Disodium selenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2Se
CAS Number:
Molecular Weight:
124.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder

SMILES string

[Na][Se][Na]

InChI

1S/2Na.Se

InChI key

PNCGYTHKTDMKME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium selenide is a useful selenylation reagent in organic synthesis. It can add a seleno group to a variety of organic substrates, such as alkenes, alkynes, and carbonyls. It is also used to synthesize organoselenium compounds, which are useful in various organic reactions.

Application

Sodium selenide can be used as a nucleophilic reagent in the reaction of ethynyl and bromoethynyl ketones to afford divinyl selenides and 1,3-enynes with high stereo- and regioselectivity. It is also used as a precursor and a stabilizing agent in the one-pot synthesis of cobalt selenide nanoparticles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service