Skip to Content
Merck
All Photos(1)

Documents

715824

Sigma-Aldrich

Diethyl (phenylsulfinylmethyl-13C)phosphonate

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC10H17O4PS
Molecular Weight:
277.28
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

liquid

mass shift

M+1

SMILES string

CCOP(=O)([13CH2]S(=O)c1ccccc1)OCC

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Licensed from Highlands Stable Isotopes Corp.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service