Skip to Content
Merck
All Photos(1)

Documents

706108

Sigma-Aldrich

Potassium 4-iodophenyltrifluoroborate

≥96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BF3IK
Molecular Weight:
309.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

form

powder

mp

141-148 °C

SMILES string

[K+].F[B-](F)(F)c1ccc(I)cc1

InChI

1S/C6H4BF3I.K/c8-7(9,10)5-1-3-6(11)4-2-5;/h1-4H;/q-1;+1

InChI key

WTXNSBOVCAXUGV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peggy E Williams et al.
Journal of the American Society for Mass Spectrometry, 29(9), 1848-1860 (2018-06-06)
In the gas phase, arylperoxyl forming reactions play a significant role in low-temperature combustion and atmospheric processing of volatile organic compounds. We have previously demonstrated the application of charge-tagged phenyl radicals to explore the outcomes of these reactions using ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service