Skip to Content
Merck
All Photos(1)

Documents

59760

Sigma-Aldrich

1-Methylimidazolium hydrogen sulfate

95%

Synonym(s):

1-Methylimidazole hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2 · H2SO4
CAS Number:
Molecular Weight:
180.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94.5% (HPLC)
95%

form

solid

impurities

≤1% water

anion traces

chloride (Cl-): ≤20 mg/kg

SMILES string

OS(O)(=O)=O.Cn1ccnc1

InChI

1S/C4H6N2.H2O4S/c1-6-3-2-5-4-6;1-5(2,3)4/h2-4H,1H3;(H2,1,2,3,4)

InChI key

TVEOIQKGZSIMNG-UHFFFAOYSA-N

Application

1-Methylimidazolium hydrogen sulfate is a Brønsted acidic ionic liquid that can be used:
  • As a component of a catalytic system along with chlorotrimethylsilane used in the preparation of 3,4-dihydropyrimidin-2(1H)-ones and hydroquinazoline-2,5-diones.
  • As a catalyst in the preparation of biologically important 2,5-dimethyl-N-substituted pyrroles.
  • As a reusable catalyst in the preparation of spiro[indoline-3′,2-quinazoline]2′,4(3H)-dione.
  • In the leaching of copper and zinc in brass dross using H2O2 as an oxidant.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>536.0 °F - (External MSDS)

Flash Point(C)

> 280 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Methylimidazolium hydrogen sulfate/chlorotrimethylsilane: an effective catalytic system for the synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones and hydroquinazoline-2, 5-diones
Kefayati H, et al.
Journal of Molecular Liquids, 172, 147-151 (2012)
An environmental friendly approach for the synthesis of spiro [indoline-3′, 2-quinazoline] 2′, 4 (3H)-dione using 1-methylimidazolium hydrogen sulfate, as reusable catalyst
Kefayati H, et al.
J. Chin. Chem. Soc., 60(10), 1197-1201 (2013)
Application of 1-methylimidazolium hydrogen sulfate ionic liquid to the oxidative leaching of copper and zinc in industrial brass dross
Energy Technology 2015, 209-215 (2015)
Daqing Li et al.
Ultrasonics sonochemistry, 20(5), 1144-1148 (2013-03-15)
Ionic liquid [HMIM]HSO4 was found to be an efficient catalyst for the synthesis of N-substituted pyrroles through the reaction of 2,5-hexanedione with amines under ultrasonic irradiation at room temperature. These reactions proceed with good yields under short reaction time. Furthermore

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service