Skip to Content
Merck
All Photos(1)

Documents

525537

Sigma-Aldrich

2-Amino-5-bromo-3-methylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7BrN2
CAS Number:
Molecular Weight:
187.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

88-95 °C (lit.)

SMILES string

Cc1cc(Br)cnc1N

InChI

1S/C6H7BrN2/c1-4-2-5(7)3-9-6(4)8/h2-3H,1H3,(H2,8,9)

InChI key

KBLGGRWUEVCNPY-UHFFFAOYSA-N

General description

2-Amino-5-bromo-3-methylpyridine can be prepared from 2-amino-3-methylpyridine, via bromination.

Application

2-Amino-5-bromo-3-methylpyridine may be used to synthesize:
  • 5-bromo-2-chloro-3-methylpyridine
  • 2-azidopyridine 1-oxide
  • 2-amino-5-bromo-3-methylpyridine 1-oxide
  • 2,5-dibromo-3-methyl pyridine
  • ethyl-3,6-dibromo-8-methylimidazo[1,2-a]pyridine-2-carboxylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-Assisted One-Pot Synthesis of Substituted 3-Bromoimidazo [1, 2-a] pyridines and Imidazoheterocycles.
Patil SM, et al.
Journal of Heterocyclic Chemistry, 51(5), 1509-1515 (2014)
2, 5-and 5, 6-Dihalonicotinic acids and their precursors.
Setliff FL.
Journal of Chemical and Engineering Data, 15(4), 590-591 (1970)
Synthesis of the grass alkaloid perlolidine through a pyridyne cyclisation reaction.
Kessar SV and Singh P.
Indian J. Chem. B, 40(11), 1129-1131 (2001)
Ring contraction of 2-azidopyridine 1-oxides and related compounds. 2-Cyano-1-hydroxypyrroles and-imidazoles.
Abramovitch RA and Cue Jr BW.
Journal of the American Chemical Society, 98(6), 1478-1486 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service