522260
PSS hydrate-Octakis(tetramethylammonium) substituted
Synonym(s):
Octakis(tetramethylammonium) pentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane-1,3,5,7,9,11,13,15-octakis(yloxide) hydrate
About This Item
Recommended Products
mp
135-143 °C (lit.)
SMILES string
O.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.C[N+](C)(C)C.[O-][Si]12O[Si]3([O-])O[Si]4([O-])O[Si]([O-])(O1)O[Si]5([O-])O[Si]([O-])(O2)O[Si]([O-])(O3)O[Si]([O-])(O4)O5
InChI
1S/8C4H12N.O20Si8.H2O/c8*1-5(2,3)4;1-21-9-22(2)12-25(5)14-23(3,10-21)16-27(7)17-24(4,11-21)15-26(6,13-22)19-28(8,18-25)20-27;/h8*1-4H3;;1H2/q8*+1;-8;
InChI key
HYWYFSRVEQSCJA-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service