Skip to Content
Merck
All Photos(1)

Documents

492175

Sigma-Aldrich

Diazinon-(diethyl-d10)

≥99 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10H11N2O3PS
CAS Number:
Molecular Weight:
314.41
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % D

Quality Level

Assay

≥98% (CP)

refractive index

n20/D 1.4978 (lit.)

bp

83-84 °C/0.002 mmHg (lit.)

density

1.152 g/mL at 25 °C

application(s)

agriculture
environmental

mass shift

M+10

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])OP(=S)(Oc1cc(C)nc(n1)C(C)C)OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3/i1D3,2D3,6D2,7D2

InChI key

FHIVAFMUCKRCQO-HXOHQZFQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diazinon-(diethyl-d10) has been used
  • In analytical methods for routine monitoring of residual pesticide.
  • As an internal standard in GC–MS quantification of pesticides in food.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoyuki Tsuchiyama et al.
Journal of chromatography. A, 1589, 122-133 (2019-01-13)
In the analysis of pesticides performed with gas chromatography, the quantitative performance of measurements can be severely compromised by phenomena known as matrix effects. In seeking a solution to the problem of matrix effects, the application of a modifier gas
Bonn Lee et al.
International journal of molecular sciences, 22(14) (2021-07-25)
A real-life environment during pregnancy involves multiple and simultaneous exposures to toxic chemicals. Perinatal exposures to toxic chemicals have been reported to exert an inhibitory effect on mouse neural development and behaviors. However, the effect of combined exposures of organophosphate
Takashi Yarita et al.
Journal of chromatography. A, 1396, 109-116 (2015-04-22)
The impact of the matrix effect in GC-MS quantification of pesticides in food using the corresponding isotope-labeled internal standards was evaluated. A spike-and-recovery study of nine target pesticides was first conducted using paste samples of corn, green soybean, carrot, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service