Skip to Content
Merck
All Photos(1)

Documents

479799

Sigma-Aldrich

Divinylphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P(CH=CH2)2
CAS Number:
Molecular Weight:
162.17
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.582 (lit.)

bp

55 °C/0.05 mmHg (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

C=CP(C=C)c1ccccc1

InChI

1S/C10H11P/c1-3-11(4-2)10-8-6-5-7-9-10/h3-9H,1-2H2

InChI key

GEKAWPIATFUQJW-UHFFFAOYSA-N

Related Categories

Application

Reactant for:
  • Synthesis of P-chiral phosphines via chiral metal template promoted asymmetric furan Diels-Alder reaction

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service