Skip to Content
Merck
All Photos(2)

Documents

463485

Sigma-Aldrich

Methyl isonipecotate

technical grade

Synonym(s):

Methyl 4-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

refractive index

n20/D 1.465 (lit.)

bp

85-90 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C1CCNCC1

InChI

1S/C7H13NO2/c1-10-7(9)6-2-4-8-5-3-6/h6,8H,2-5H2,1H3

InChI key

RZVWBASHHLFBJF-UHFFFAOYSA-N

General description

Methyl isonipecotate can be obtained by the reduction of methyl isonicotinate in methanol with ruthenium at 100-150°C.

Application

Methyl isonipecotate may be used for the synthesis of a series of potent non-urea soluble epoxide hydrolase (sEH) inhibitors via high throughput screens.
Precipitate may form upon standing which should not affect application.
Reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation

Reactant for synthesis of:
Antitubercular agents
Anti-HIV-1 agents
Aminopyrazine inhibitors
Orally available naphthyridine protein kinase D inhibitors
A Scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Reaction of Calcium with Pyridine and Its Methyl Derivatives1.
Utke AR and Sanderson RT.
The Journal of Organic Chemistry, 28(12), 3582-3584 (1963)
Yuli Xie et al.
Bioorganic & medicinal chemistry letters, 19(8), 2354-2359 (2009-03-24)
Soluble epoxide hydrolase (sEH) is a novel target for the treatment of hypertension and vascular inflammation. A new class of potent non-urea sEH inhibitors was identified via high throughput screening (HTS) and chemical modification. IC(50)s of the most potent compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service