Skip to Content
Merck
All Photos(1)

Documents

462551

Sigma-Aldrich

Titanium(IV) tert-butoxide

deposition grade

Synonym(s):

Tetra-tert-butyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti[OC(CH3)3]4
CAS Number:
Molecular Weight:
340.32
Beilstein:
3681801
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Quality Level

Assay

98.5-101.5% (gravimetric)

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

density

0.881 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)O[Ti](OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

InChI

1S/4C4H9O.Ti/c4*1-4(2,3)5;/h4*1-3H3;/q4*-1;+4

InChI key

GRWPYGBKJYICOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

117.0 °F - closed cup

Flash Point(C)

47.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjuan Ding et al.
Membranes, 10(3) (2020-03-19)
Dense and good catalytic performance TS-1 zeolite membranes were rapidly prepared on porous mullite support by secondary hydrothermal synthesis. The properties of seed crystals were very important for the preparation of high-catalytic performance TS-1 zeolite membranes. Influences of seed crystals
Marina Matos et al.
Materials (Basel, Switzerland), 12(2) (2019-01-24)
Poly(ether ester)s (PEEs) represent a promising class of segmented co-polymers, nevertheless the synthesis of PEEs based on renewable 2,5-furandicarboxylic acid (FDCA) is still scarce. In this context, a series of poly(1,4-butylene 2,5-furandicarboxylate)-co-poly(poly(propylene oxide) 2,5-furandicarboxylate) co-polyesters with different composition of stiff
Changdeuck Bae et al.
Scientific reports, 5, 9339-9339 (2015-03-24)
Rational synthesis of coordinated spherical colloids is reported by site-selective growth of secondary hemispherical patches on primary spherical particles with quasi-defined coordination numbers and positions. We clarify the importance of mass transport phenomena on the site-specific secondary nucleation/growth in nanoparticulate

Articles

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service