Recommended Products
Quality Level
Assay
95%
form
solid
optical activity
[α]25/D +11.0°, c = 1 in methanol
reaction suitability
reaction type: solution phase peptide synthesis
mp
212-217 °C (lit.)
application(s)
peptide synthesis
SMILES string
Cl[H].C[C@H](N)C(N)=O
InChI
1S/C3H8N2O.ClH/c1-2(4)3(5)6;/h2H,4H2,1H3,(H2,5,6);1H/t2-;/m0./s1
InChI key
FIAINKIUSZGVGX-DKWTVANSSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service