Skip to Content
Merck
All Photos(1)

Key Documents

439738

Sigma-Aldrich

Manganese(II) iodide

anhydrous, 99.99% trace metals basis

Synonym(s):

Manganese diiodide, Manganese iodide, Manganous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnI2
CAS Number:
Molecular Weight:
308.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder or crystals (or flakes)

impurities

≤150.0 ppm Trace Metal Analysis

mp

80 °C (dec.) (lit.)

density

5.01 g/mL at 25 °C (lit.)

SMILES string

I[Mn]I

InChI

1S/2HI.Mn/h2*1H;/q;;+2/p-2

InChI key

QWYFOIJABGVEFP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service