421650
(2S-cis)-(−)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid
97%
Synonym(s):
(2S,5S)-3,6-Dioxo-5-(phenylmethyl)-2-piperazineacetic acid, Cycloaspartylphenylalanine
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder
optical activity
[α]20/D −9.0°, c = 0.9 in acetic acid
mp
270-272 °C (lit.)
functional group
carboxylic acid
phenyl
SMILES string
OC(=O)C[C@@H]1NC(=O)[C@H](Cc2ccccc2)NC1=O
InChI
1S/C13H14N2O4/c16-11(17)7-10-13(19)14-9(12(18)15-10)6-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2,(H,14,19)(H,15,18)(H,16,17)/t9-,10-/m0/s1
InChI key
VNHJXYUDIBQDDX-UWVGGRQHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- T cell lymphomatoid contact dermatitis: a challenging case and review of the literature.: This review includes discussions on 2-Methyl-4-isothiazolin-3-one-induced contact dermatitis, providing valuable information for dermatological applications and safety assessments (Knackstedt and Zug, 2015).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service