All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
optical activity
[α]20/D +12°, c = 2 in chloroform
refractive index
n20/D 1.455 (lit.)
bp
55 °C/18 mmHg (lit.)
density
0.98 g/mL at 25 °C (lit.)
functional group
amine
ether
SMILES string
NC[C@@H]1CCCO1
InChI
1S/C5H11NO/c6-4-5-2-1-3-7-5/h5H,1-4,6H2/t5-/m0/s1
InChI key
YNOGYQAEJGADFJ-YFKPBYRVSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service