407968
Ethylene glycol dicyclopentenyl ether acrylate
contains 700 ppm monomethyl ether hydroquinone as inhibitor
Synonym(s):
2-(Dicyclopenten-6-yloxy)ethyl acrylate, Dicyclopentyloxyethyl acrylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
contains
700 ppm monomethyl ether hydroquinone as inhibitor
refractive index
n20/D 1.501 (lit.)
density
1.085 g/mL at 25 °C (lit.)
SMILES string
C=CC(=O)OCCOC1CC2CC1C3C=CCC23
InChI
1S/C15H20O3/c1-2-15(16)18-7-6-17-14-9-10-8-13(14)12-5-3-4-11(10)12/h2-4,10-14H,1,5-9H2
InChI key
RSVDRWTUCMTKBV-UHFFFAOYSA-N
Related Categories
General description
Ethylene glycol dicyclopentenyl ether acrylate (EGDPEA) is a versatile acrylate monomer characterized by its dicyclopentenyl ether structure, which imparts unique properties such as enhanced thermal stability, UV resistance, and mechanical strength. Its acrylate functionality allows for rapid polymerization and easy incorporation into various polymer matrices, making it suitable for a wide range of applications in polymer synthesis, drug delivery and biomedical devices.
Application
Ethylene glycol dicyclopentenyl ether acrylate can be used:
- As a base monomer to prepare ink-jet 3D printing formulation to fabricate biofilm resistant medical devices such as stents, orthopedic implants, and dental materials. Its mechanical strength and biocompatibility ensure that these devices can withstand physiological conditions while minimizing the risk of inflammation or infection.
- As a precursor to synthesize biocompatible acrylate-based 3D scaffolds for tissue repair, due to its high stem cell attachment properties.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
201.2 °F - closed cup
Flash Point(C)
94 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service