Skip to Content
Merck
All Photos(2)

Documents

386561

Sigma-Aldrich

Ethyl methylphosphonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(O)(OH)OC2H5
CAS Number:
Molecular Weight:
124.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.426 (lit.)

density

1.172 g/mL at 25 °C (lit.)

SMILES string

CCOP(C)(O)=O

InChI

1S/C3H9O3P/c1-3-6-7(2,4)5/h3H2,1-2H3,(H,4,5)

InChI key

UFZOPKFMKMAWLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Kammer et al.
Biosensors & bioelectronics, 131, 119-127 (2019-03-04)
Organophosphorus compounds (OPs) continue to represent a significant chemical threat to humans due to exposures from their use as weapons, their potential storage hazards, and from their continued use agriculturally. Existing methods for detection include ELISA and mass spectrometry. The
Boris Sarvin et al.
Journal of chromatography. A, 1597, 214-219 (2019-04-24)
In this research an accumulation of nerve agent markers in garden cress (Lepidium sativum) as a model plant object was studied using LC-QTOF hybrid system. For the determination of methylphosphonic acid and alkyl methylphosphonates, which are specific markers of sarin
Dalton T Snyder et al.
Journal of the American Society for Mass Spectrometry, 29(7), 1345-1354 (2018-03-11)
Methodology for performing precursor and neutral loss scans in an RF scanning linear quadrupole ion trap is described and compared to the unconventional ac frequency scan technique. In the RF scanning variant, precursor ions are mass selectively excited by a
Alan C Samuels et al.
Analytical chemistry, 78(2), 408-415 (2006-01-18)
Diffuse reflection data are presented for ethyl methylphosphonate in a fine Utah dirt sample as a model system for organophosphate-contaminated soil. The data revealed a chemometric artifact when the spectra were represented in Kubelka-Munk units that manifests as a linear
Sophie Le Moullec et al.
Journal of chromatography. A, 1108(1), 7-13 (2006-02-03)
The analysis of alkyl alkylphosphonic acids, the degradation products of V and G nerve agents as VX, Sarin or Soman, is an important task for the verification of compliance to the Chemical Weapons Convention. The detection of these contaminants at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service