Skip to Content
Merck
All Photos(2)

Key Documents

386421

Sigma-Aldrich

N-tert-Butylmaleimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO2
CAS Number:
Molecular Weight:
153.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

refractive index

n20/D 1.477 (lit.)

bp

189 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

functional group

imide
maleimide

SMILES string

CC(C)(C)N1C(=O)C=CC1=O

InChI

1S/C8H11NO2/c1-8(2,3)9-6(10)4-5-7(9)11/h4-5H,1-3H3

InChI key

YEKDUBMGZZTUDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Zhao et al.
Analytica chimica acta, 1037, 274-280 (2018-10-08)
Ratiometric quantitation is used in mass spectrometry to account for variations in ionization efficiencies due to heterogenous sample matrixes. Isotopes are most commonly used to achieve ratiometric quantitation because of their ability to co-elute chromatographically with each other and to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service