Skip to Content
Merck
All Photos(2)

Key Documents

278386

Sigma-Aldrich

2,4,7,9-Tetramethyl-5-decyne-4,7-diol, mixture of (±) and meso

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)(OH)C≡CC(CH3)(OH)CH2CH(CH3)2
CAS Number:
Molecular Weight:
226.36
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

bp

255 °C (lit.)

mp

42-44 °C (lit.)

HLB

4

SMILES string

CC(C)CC(C)(O)C#CC(C)(O)CC(C)C

InChI

1S/C14H26O2/c1-11(2)9-13(5,15)7-8-14(6,16)10-12(3)4/h11-12,15-16H,9-10H2,1-6H3

InChI key

LXOFYPKXCSULTL-UHFFFAOYSA-N

General description

2, 4, 7, 9-tetramethyldec-5-yne-4,7-diol is a well-known acrylic adhesive. It has amphiphilic characteristics. It is used a dopant in a heterojunction solar cell, it assists in the formation of a homogeneous interfacial layer over the photoactive layer. Doping results in decreasing the absorption of photons in the underneath photoactive layer, thereby in enhancing the photocurrent density.

Application

2, 4, 7, 9-tetramethyldec-5-yne-4,7-diol may be used to dope poly[9,9-bis(6′-(diethanolamino)hexyl)-fluorene],that acts as an interfacial layer in a heterojunction solar cell.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Canellas et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(10), 1721-1729 (2017-03-24)
The suitability of an acrylic adhesive used on food packaging was studied. Six potential migrants were identified using GC-MS and UPLC-QTOF. Five compounds were intentionally added (2-butoxyethanol and 2,4,7,9-tetramethyl-5-decyne-4,7-diol 10 (TMDD) and TMDD ethoxylates). One of the compounds identified as
Sergio Rodríguez et al.
Journal of environmental management, 255, 109926-109926 (2020-02-18)
Treatment of polluted wastewaters from industrial activities has become a source of major concern for the environment. In this work, real wastewater from a physico-chemical (WWFQ) treatment was tested through different oxidation technologies: Fenton and Fenton-like reagent and persulfate activated
Enhanced performance in bulk heterojunction solar cells with alkylidene fluorene donor by introducing modified PFN-OH/Al bilayer cathode
Heo SW, et al.
Royal Society of Chemistry Advances, 4, 6776-6781 (2014)
Partition and diffusion of volatile compounds from acrylic adhesives used for food packaging multilayers manufacturing
Cabellas E, et al.
Journal of Materials Chemistry, 20, 5100-5109 (2010)
Betsy T Kren et al.
Breast cancer research : BCR, 17, 19-19 (2015-04-04)
Targeted therapies for aggressive breast cancers like triple negative breast cancer (TNBC) are needed. The use of small interfering RNAs (siRNAs) to disable expression of survival genes provides a tool for killing these cancer cells. Cyclin dependent kinase 11 (CDK11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service