Skip to Content
Merck
All Photos(1)

Documents

275883

Sigma-Aldrich

3-Chloro-2-nitrobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
Beilstein:
2109828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

237-239 °C (lit.)

SMILES string

OC(=O)c1cccc(Cl)c1[N+]([O-])=O

InChI

1S/C7H4ClNO4/c8-5-3-1-2-4(7(10)11)6(5)9(12)13/h1-3H,(H,10,11)

InChI key

VCHSXYHBMFKRBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen-bonded structures of isomeric compounds of 3-chloro-2-nitrobenzoic acid with quinoline have been investigated.

Application

3-Chloro-2-nitrobenzoic acid has been used in the preparation of:
  • 2-amino-3-chlorobenzonitrile
  • 3-chloro-2-nitrobenzaldehyde

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuma Gotoh et al.
Acta crystallographica. Section C, Crystal structure communications, 65(Pt 10), o534-o538 (2009-10-07)
The structures of four isomeric compounds, all C7H4ClNO4.C9H7N, of quinoline with chloro- and nitro-substituted benzoic acid, namely, 2-chloro-5-nitrobenzoic acid-quinoline (1/1), (I), 3-chloro-2-nitrobenzoic acid-quinoline (1/1), (II), 4-chloro-2-nitrobenzoic acid-quinoline (1/1), (III), and 5-chloro-2-nitrobenzoic acid-quinoline (1/1), (IV), have been determined at 185 K.
The synthesis and in vitro acetylcholinesterase and butyrylcholinesterase inhibitory activity of tacrine (Cognex?) derivaties.
Gregor VE, et al.
Bioorganic & Medicinal Chemistry Letters, 2(8), 861-864 (1992)
Tadashi Kataoka et al.
Bioorganic & medicinal chemistry, 12(9), 2397-2407 (2004-04-15)
Condensation of nitrobenzaldehydes 3 and alpha-[o-(p-methoxybenzylthio)benzoyl] sulfoxide 4 gave alpha-sulfinyl enones 5. Treatment of 5 with formic acid caused cyclization followed by debenzylation to afford 3-(methylsulfinyl)thioflavanones 6. Double-bond formation with elimination of methanesulfenic acid was performed by refluxing 6 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service