Skip to Content
Merck
All Photos(3)

Key Documents

203521

Sigma-Aldrich

Lanthanum(III) chloride heptahydrate

99.999% trace metals basis

Synonym(s):

Lanthanum trichloride heptahydrate, Lanthanum(3+) trichloride heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3 · 7H2O
CAS Number:
Molecular Weight:
371.37
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤15.0 ppm Trace Rare Earth Analysis

mp

91 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.O.O.Cl[La](Cl)Cl

InChI

1S/3ClH.La.7H2O/h3*1H;;7*1H2/q;;;+3;;;;;;;/p-3

InChI key

FDFPDGIMPRFRJP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum(III)chloride heptahydrate is a mild, inexpensive, and moisture-stable heterogeneouscatalyst widely used in several organic transformations.

Application

Lanthanum(III) chloride heptahydrate can be used as an efficient catalyst for :
  • Solvent-free preparation of substituted alkenes via Knoevenagel condensation.
  • One-pot synthesis of 3,4-dihydropyrimidin-2(1H)-ones.
It can be used as a precursor to synthesize La/V co-doped TiO2 nanoparticles through the sol-gel method. La co-doped TiO2 nanopowder show enhanced photocatalytic activity.

Biochem/physiol Actions

Calcium channel blocker

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuta Kawarasaki et al.
Journal of insect physiology, 120, 103996-103996 (2019-12-15)
Rapid cold-hardening (RCH) is a type of phenotypic plasticity that promotes a swift improvement of cold tolerance in insects. A brief exposure to mild cold dramatically increases insect survival to a subsequent cold exposure that would be lethal otherwise. In
Anna Citta et al.
Journal of inorganic biochemistry, 117, 18-24 (2012-10-20)
Lanthanum chloride (LaCl(3)) is a good inhibitor of both the cytosolic and mitochondrial thioredoxin reductase with IC(50) values of 1.75 and 7.46 μM, respectively. On the contrary, the related enzyme glutathione reductase is not inhibited by lanthanum ions even at
Tawnya G Flick et al.
Journal of the American Society for Mass Spectrometry, 23(11), 1885-1895 (2012-09-06)
Addition of 1.0 mM LaCl(3) to aqueous ammonium acetate solutions containing proteins in their folded native forms can result in a significant increase in the molecular ion charging obtained with electrospray ionization as a result of cation adduction. In combination
T Wu et al.
American journal of physiology. Heart and circulatory physiology, 301(1), H69-H78 (2011-04-05)
The cochlear lateral wall generates the endocochlear potential (EP), which creates a driving force for the hair cell transduction current and is essential for normal hearing. Blood flow at the cochlear lateral wall is critically important for maintaining the EP.
Long Zhang et al.
Journal of environmental sciences (China), 24(5), 940-948 (2012-08-17)
The effect of calcium chloride (CaCl2) on rice seedling growth under cadmium chloride (CdCl2) stress, as well as the possible role of endogenous nitric oxide (NO) in this process, was studied. The growth of rice seedlings was seriously inhibited by

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service