Skip to Content
Merck
All Photos(1)

Documents

158348

Sigma-Aldrich

4-(Benzyloxy)phenol

98%

Synonym(s):

4-(Phenylmethoxy)phenol, Hydroquinone monobenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4OH
CAS Number:
Molecular Weight:
200.23
Beilstein:
1958305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

119-120 °C (lit.)

SMILES string

Oc1ccc(OCc2ccccc2)cc1

InChI

1S/C13H12O2/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9,14H,10H2

InChI key

VYQNWZOUAUKGHI-UHFFFAOYSA-N

Gene Information

human ... TYR(7299)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

4-(Benzyloxy)phenol was used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It was also used in the preparation of series of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, spectral properties, biological activity and application of new 4-(benzyloxy) phenol derived azo dyes for polyester fiber dyeing.
Yousefi H, et al.
Journal of Molecular Liquids, 180, 51-58 (2013)
K Urabe et al.
Seminars in cutaneous medicine and surgery, 16(1), 81-85 (1997-03-01)
The dyschromatoses are a group of disorders characterized by the presence of both hyperpigmented and hypopigmented macules, many of which are small in size and irregular in shape. There are two major forms-dyschromatosis symmetrica hereditaria (DSH) and dyschromatosis universalis hereditaria
K M AlGhamdi et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 25(7), 749-757 (2010-11-09)
If vitiligo involves most of the body, it might be easier to depigment the normal remaining skin rather than to attempt repigmentation. We reviewed the literature to date regarding available therapies for depigmenting the normal skin in vitiligo universalis. Our
L A Rheins et al.
Journal of immunology (Baltimore, Md. : 1950), 136(3), 867-876 (1986-02-01)
The epidermis on the backs or ears of DBA/2 mice treated for 7 days with a 20% concentration of monobenzyl ether of hydroquinone (MBEH) had a significantly greater population density of ATPase- and Ia-positive cells compared with control mice treated
W C Quevedo et al.
Pigment cell research, 3(2), 71-79 (1990-03-01)
Topical applications of monobenzylether of hydroquinone (MBEH) or intraperitoneal injections of phenol induced graying of hair in eumelanic mice but had little effect on hair color in pheomelanic mice. Amcinonide, an anti-inflammatory agent, elicited whitening of a few hairs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service