Skip to Content
Merck
All Photos(1)

Key Documents

156841

Sigma-Aldrich

(+)-Diethyl L-tartrate

≥99%

Synonym(s):

L-(+)-Tartaric acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CO2C2H5]2
CAS Number:
Molecular Weight:
206.19
Beilstein:
1727145
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

optical activity

[α]20/D +8.5°, neat

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.446 (lit.)

bp

280 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CCOC(=O)[C@H](O)[C@@H](O)C(=O)OCC

InChI

1S/C8H14O6/c1-3-13-7(11)5(9)6(10)8(12)14-4-2/h5-6,9-10H,3-4H2,1-2H3/t5-,6-/m1/s1

InChI key

YSAVZVORKRDODB-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Made from natural tartaric acid

Application

(+)-Diethyl L-tartrate can be used in the synthesis of biologically active compounds such as (+)-altholactone, (-)-aspicilin, (+)-monomorine I and (+)-(1R,2R,3S,6S)-3,6-di-O-methyl conduritol-E.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An enantioselective total synthesis of (+)-altholactone from diethyl L-tartrate.
Somfai P.
Tetrahedron, 50(38), 11315-11320 (1994)
A new and concise synthetic route to an enantiopure (+)-conduritol-E derivative from diethyl l-tartrate.
Lee WW and Chang S.
Tetrahedron Asymmetry, 10(23), 4473-4475 (1999)
Enantioselective total synthesis of (+)-monomorine I.
Yamazaki N and Kibayashi C.
Tetrahedron Letters, 29(45), 5767-5768 (1988)
Stereocontrolled total synthesis of the macrocyclic lactone (-)-aspicilin.
Waanders PP, et al.
Tetrahedron Letters, 28(21), 2409-2412 (1987)
Kimberly A Kahle et al.
Electrophoresis, 28(15), 2644-2657 (2007-06-29)
In this second study on dual-chirality microemulsions containing a chiral surfactant and a chiral oil, a less hydrophobic and lower interfacial tension chiral oil, diethyl tartrate, is employed (Part 1, Foley, J. P. et al.., Electrophoresis, DOI: 10.1002/elps.200600551). Six stereochemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service