Skip to Content
Merck
All Photos(1)

Key Documents

131563

Sigma-Aldrich

Propyltriphenylphosphonium bromide

98%

Synonym(s):

NSC 50539

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2P(C6H5)3Br
CAS Number:
Molecular Weight:
385.28
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

235-238 °C (lit.)

SMILES string

[Br-].CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H22P.BrH/c1-2-18-22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1

InChI key

XMQSELBBYSAURN-UHFFFAOYSA-M

Application

Catalyst for the carboxylation of styrene oxide

Reactant involved in:
  • Epoxidation of arylalkenes
  • Boration of allylic epoxides
  • Wittig reactions
  • Selective dimerization of α-olefins
  • Synthesis of indoles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service