Merck
  • Protective effects of piperine on the retina of mice with streptozotocin-induced diabetes by suppressing HIF-1/VEGFA pathway and promoting PEDF expression.
Protective effects of piperine on the retina of mice with streptozotocin-induced diabetes by suppressing HIF-1/VEGFA pathway and promoting PEDF expression.
International journal of ophthalmology (2021-05-21)
Pu Zhang, Yan-Dan Zhou, Yao Tan, Ling Gao
ABSTRACT
To evaluate the protective mechanisms of piperine in the retina of mice with streptozotocin-induced diabetes. In experiments in vitro, stimulation by chemical hypoxia was established in ARPE-19 cells. Then, the expression of hypoxia-inducible factor-1α (HIF-1α), vascular endothelial growth factor A (VEGFA), and pigment epithelium-derived factor (PEDF) was assessed at the mRNA and protein levels. In experiments in vivo, diabetes mellitus was established by intraperitoneally injecting 150 mg/kg streptozotocin once. After 3wk of the onset of diabetes, 15 mg/kg piperine was intraperitoneally injected once daily for 1 or 3wk. Then, the retinal morphology and mRNA and protein expression were assessed. In hypoxia, 1-100 µmol/L piperine significantly decreased the expression of VEGFA mRNA and increased the expression of PEDF mRNA without affecting HIF-1α mRNA. Meanwhile, 100 µmol/L piperine substantially decreased the protein level of VEGFA and increased the protein level of PEDF. The HIF-1α protein level was also hampered by piperine. In the diabetic retina of mice, the morphological damage was alleviated by piperine. Likewise, the retinal vascular leakage was substantially decreased by piperine. Further, the protein levels of HIF-1α and VEGFA were significantly reduced by piperine. Moreover, the level of the antiangiogenic factor of PEDF dramatically increased by piperine. Piperine may exert protective effects on the retina of mice with diabetes via regulating the pro-antiangiogenic homeostasis composed of HIF-1/VEGFA and PEDF.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-PEDF Antibody, Upstate®, from rabbit