![Paclitaxel Impurity C certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/172/551/5ff8acef-511d-48c1-b591-19cb563f38bb/640/5ff8acef-511d-48c1-b591-19cb563f38bb.jpg)
Paclitaxel Impurity C
![Paclitaxel Impurity C certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/172/551/5ff8acef-511d-48c1-b591-19cb563f38bb/640/5ff8acef-511d-48c1-b591-19cb563f38bb.jpg)
Synonym(s):
N-Debenzoyl-N-hexanoylpaclitaxel
Empirical Formula (Hill Notation):
C46H57NO14
CAS Number:
Molecular Weight:
847.94
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2